APO945299098 Apollo Scientific Ltd

2-Fluoro-3-nitrophenol

  • MDL number MFCD11042443
  • Cas Number : 179816-26-3
  • Molecular Weight : 157.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-3-nitrophenol
Inchi Code 1S/C6H4FNO3/c7-6-4(8(10)11)2-1-3-5(6)9/h1-3,9H
Inchi Key QZOGSUCWBIPKDC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-nitrophenol

Safety Information

signalword Danger
pcodes P260;P280;P301+P312;P302+P352;P304+P340;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-nitrophenol
pictograms GHS05;GHS07;GHS08
hstatements H302;H317;H318;H335;H373;H412
Availability details for
Quantity
  • Availability
  • Add to Cart