APO945299100 Apollo Scientific Ltd

Methyl 3-bromo-4,5-difluorobenzoate

  • MDL number MFCD20483155
  • Cas Number : 1244642-70-3
  • Molecular Weight : 251.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-bromo-45-difluorobenzoate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-bromo-45-difluorobenzoate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart