APO945299103 Apollo Scientific Ltd

Trimethylsilyl 2-chloro-2,2-difluoroacetate

  • MDL number MFCD30723300
  • Cas Number : 1700654-90-5
  • Molecular Weight : 202.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym TCDA
Storage temp. Store at 2-8
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/trimethylsilyl-2-chloro-22-difluoroacetate

Safety Information

signalword Danger
pcodes P280;P301+P330+P331;P308+P313
MSDS https://store.apolloscientific.co.uk/product/trimethylsilyl-2-chloro-22-difluoroacetate
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart