APO945299105 Apollo Scientific Ltd

7-Bromo-6-fluoro-2H-3,1-benzoxazine-2,4(1H)-dione

  • MDL number MFCD30723301
  • Cas Number : 1887002-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/7-bromo-6-fluoro-2h-31-benzoxazine-241h-dione

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/7-bromo-6-fluoro-2h-31-benzoxazine-241h-dione
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart