APO945299106 Apollo Scientific Ltd

2,6-Difluoro-3-(trifluoromethyl)benzyl alcohol

  • MDL number MFCD30723302
  • Cas Number : 1800231-22-4
  • Molecular Weight : 212.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-3-trifluoromethylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-3-trifluoromethylbenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart