APO945299107 Apollo Scientific Ltd

4,5-Difluoro-2-iodotoluene

  • MDL number MFCD12547764
  • Cas Number : 1208077-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8°C
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/45-difluoro-2-iodotoluene

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/45-difluoro-2-iodotoluene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart