APO945299108 Apollo Scientific Ltd

3-(Trifluoromethylsulphonyl)benzoic acid

  • MDL number MFCD00766763
  • Cas Number : 952-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-trifluoromethylsulphonylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-trifluoromethylsulphonylbenzoic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart