APO945299114 Apollo Scientific Ltd

3-(Cyanomethyl)-5-fluorobenzeneboronic acid

  • MDL number MFCD27935531
  • Cas Number : 1460307-63-4
  • Molecular Weight : 178.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-cyanomethyl-5-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-cyanomethyl-5-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart