APO945299116 Apollo Scientific Ltd

2-Bromo-3-hydroxy-6-(trifluoromethyl)pyridine

  • MDL number MFCD15478054
  • Cas Number : 1211526-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-6-(trifluoromethyl)pyridin-3-ol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=105390

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=105390
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart