APO945299154 Apollo Scientific Ltd

Xenon difluoride

  • MDL number MFCD00040538
  • Cas Number : 13709-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129
Purity 99.90
Density 4.32
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/xenon-difluoride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/xenon-difluoride
pictograms GHS03;GHS05;GHS06
hstatements H272;H301;H314;H330
Availability details for
Quantity
  • Availability
  • Add to Cart