APO946845005 Apollo Scientific Ltd

4-Iodoindoline-2,3-dione

  • MDL number MFCD03427300
  • Cas Number : 20780-75-0
  • Molecular Weight : 273.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-iodoindoline-23-dione

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/4-iodoindoline-23-dione
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart