APO948334955 Apollo Scientific Ltd

2-Methoxy-3-methylbenzyl alcohol

  • MDL number MFCD12031989
  • Cas Number : 74090-48-5
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methoxy-3-methylphenyl)methanol
Inchi Code 1S/C9H12O2/c1-7-4-3-5-8(6-10)9(7)11-2/h3-5,10H,6H2,1-2H3
Inchi Key ARAAMZJQNNALGN-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-3-methylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-3-methylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart