APO948335004 Apollo Scientific Ltd

6-Bromo-2-fluoro-3-iodobenzoic acid

  • MDL number MFCD24848699
  • Cas Number : 217816-53-0
  • Molecular Weight : 344.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-2-fluoro-3-iodobenzoic acid
Inchi Code 1S/C7H3BrFIO2/c8-3-1-2-4(10)6(9)5(3)7(11)12/h1-2H,(H,11,12)
Inchi Key FTGARIJZXXCFHE-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-2-fluoro-3-iodobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-2-fluoro-3-iodobenzoic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart