APO948335032 Apollo Scientific Ltd

3-Bromo-2-iodoaniline

  • MDL number MFCD16622211
  • Cas Number : 150908-00-2
  • Molecular Weight : 297.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-iodoaniline

Safety Information

signalword Danger
pcodes P260;P280;P301+P310
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-iodoaniline
pictograms GHS06
hstatements H301;H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart