APO948335049 Apollo Scientific Ltd

Methyl 5-amino-2-bromobenzoate

  • MDL number MFCD00085568
  • Cas Number : 6942-37-6
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-amino-2-bromobenzoate

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-amino-2-bromobenzoate
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart