APO959408659 Apollo Scientific Ltd

4-Amino-3,5-dichloropyridazine

  • MDL number MFCD01647251
  • Cas Number : 53180-76-0
  • Molecular Weight : 163.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3,5-Dichloropyridazin-4-amine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-35-dichloropyridazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-35-dichloropyridazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart