APO959408675 Apollo Scientific Ltd

Icosanedioic acid

  • MDL number MFCD00059627
  • Cas Number : 2424-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127
Synonym 1,18-Octadecanedicarboxylic acid, Eicosanedioic acid
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 234
Product Link http://www.apolloscientific.co.uk/display_item.php?id=107303

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=107303
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart