APO959408683 Apollo Scientific Ltd

3,5-Difluoro-4-(hydroxymethyl)phenol

  • MDL number MFCD11040173
  • Cas Number : 438049-36-6
  • Molecular Weight : 160.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 160
Iupac Name 3,5-difluoro-4-(hydroxymethyl)phenol
Inchi Code 1S/C7H6F2O2/c8-6-1-4(11)2-7(9)5(6)3-10/h1-2,10-11H,3H2
Inchi Key RQMJWCOEDWMJLS-UHFFFAOYSA-N

Description

Synonym 2,6-Difluoro-4-hydroxybenzyl alcohol, (2,6-Difluoro-4-hydroxyphenyl)methanol
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 160
Product Link https://store.apolloscientific.co.uk/product/35-difluoro-4-hydroxymethylphenol

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/35-difluoro-4-hydroxymethylphenol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart