APO959408685 Apollo Scientific Ltd

3-Fluoro-5-nitrotoluene

  • MDL number MFCD06797943
  • Cas Number : 499-08-1
  • Molecular Weight : 155.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-fluoro-3-methyl-5-nitrobenzene
Inchi Code 1S/C7H6FNO2/c1-5-2-6(8)4-7(3-5)9(10)11/h2-4H,1H3
Inchi Key IPMURRZVEWZEPP-UHFFFAOYSA-N

Description

Synonym 1-Fluoro-3-methyl-5-nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-nitrotoluene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-nitrotoluene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart