APO959408697 Apollo Scientific Ltd

Methyl 3-fluoro-4-(methylsulphonyl)benzoate

  • MDL number MFCD15143295
  • Cas Number : 1215074-49-9
  • Molecular Weight : 232.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-fluoro-4-(methylsulfonyl)benzoate
Inchi Code 1S/C9H9FO4S/c1-14-9(11)6-3-4-8(7(10)5-6)15(2,12)13/h3-5H,1-2H3
Inchi Key ACDGZYOVJKPTRE-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-fluoro-4-methylsulphonylbenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-fluoro-4-methylsulphonylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart