APOH03871BD0 Apollo Scientific Ltd

3-(Dimethylamino)-1-(2-furyl)prop-2-en-1-one

  • MDL number MFCD00121186
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-dimethylamino-1-2-furylprop-2-en-1-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-dimethylamino-1-2-furylprop-2-en-1-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart