APOH03871BE1 Apollo Scientific Ltd

Ethyl 1-(5-amino-1,3,4-thiadiazol-2-yl)piperidine-3-carboxylate 1g pack

  • MDL number MFCD29060042
  • Cas Number : 2109229-22-1
  • Molecular Weight : 256.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-1-5-amino-134-thiadiazol-2-ylpiperidine-3-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart