APOH03871BF0 Apollo Scientific Ltd

Ethyl 3-bromo-4-hydroxy-5-nitrobenzoate

  • Cas Number : 141778-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=111296

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=111296
Availability details for
Quantity
  • Availability
  • Add to Cart