APOH03871BF6 Apollo Scientific Ltd

2-(Chloromethyl)-5-methyl-1,3-benzoxazole

  • MDL number MFCD07366537
  • Cas Number : 41014-44-2
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloromethyl-5-methyl-13-benzoxazole

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/2-chloromethyl-5-methyl-13-benzoxazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart