APOH03871BFF Apollo Scientific Ltd

Methyl 3-chloro-1H-indazole-7-carboxylate

  • Cas Number : 1337881-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=111319

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=111319
Availability details for
Quantity
  • Availability
  • Add to Cart