APOH03871C0E Apollo Scientific Ltd

7-Methylimidazo[1,2-a]pyridine-8-carbonitrile hydrochloride

  • Cas Number : 2108781-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=111340

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=111340
Availability details for
Quantity
  • Availability
  • Add to Cart