APOH03871C2F Apollo Scientific Ltd

3,7-Dichloroimidazo[1,2-a]pyridine

  • Cas Number : 1019026-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=111399

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=111399
Availability details for
Quantity
  • Availability
  • Add to Cart