APOH03871C40 Apollo Scientific Ltd

1-Methyl-1H-pyrrole-2-carbonitrile

  • MDL number MFCD02693721
  • Cas Number : 34884-10-1
  • Molecular Weight : 106.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.03
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-methyl-1h-pyrrole-2-carbonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-1h-pyrrole-2-carbonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart