APOH03871C42 Apollo Scientific Ltd

(6-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid

  • MDL number MFCD06739269
  • Cas Number : 59128-15-3
  • Molecular Weight : 255.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromoimidazo12-apyridin-2-ylacetic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromoimidazo12-apyridin-2-ylacetic-acid
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart