APOH03871C50 Apollo Scientific Ltd

4-Bromo-1,3-dimethyl-1H-pyrazol-5-amine hydrobromide

  • Cas Number : 1431965-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=111440

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=111440
Availability details for
Quantity
  • Availability
  • Add to Cart