APOH03871CBD Apollo Scientific Ltd

2-(2-Chloro-3,4-dimethoxyphenyl)acetic acid

  • MDL number MFCD21606658
  • Cas Number : 6834-51-1
  • Molecular Weight : 230.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chloro-3,4-dimethoxyphenyl)acetic acid
Inchi Code 1S/C10H11ClO4/c1-14-7-4-3-6(5-8(12)13)9(11)10(7)15-2/h3-4H,5H2,1-2H3,(H,12,13)
Inchi Key RAMSELFCQDNGKU-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-2-chloro-34-dimethoxyphenylacetic-acid

Safety Information

signalword TBC
pcodes TBC
MSDS https://store.apolloscientific.co.uk/product/2-2-chloro-34-dimethoxyphenylacetic-acid
pictograms TBC
hstatements TBC
Availability details for
Quantity
  • Availability
  • Add to Cart