APOH03871CC8 Apollo Scientific Ltd

Ethyl 4-iodo-5-methylisoxazole-3-carboxylate

  • MDL number MFCD28992249
  • Cas Number : 1356600-24-2
  • Molecular Weight : 281.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-iodo-5-methylisoxazole-3-carboxylate
Inchi Code 1S/C7H8INO3/c1-3-11-7(10)6-5(8)4(2)12-9-6/h3H2,1-2H3
Inchi Key CIAFQPZJFZGWNG-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-4-iodo-5-methylisoxazole-3-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-4-iodo-5-methylisoxazole-3-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart