APOH03871D0E Apollo Scientific Ltd

8-Chloro-4H-thieno[3,2-c]thiochromene-2-carboxylic acid

  • Cas Number : 255395-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=111667

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=111667
Availability details for
Quantity
  • Availability
  • Add to Cart