APOH03871D72 Apollo Scientific Ltd

2,4-Difluoro-5-methylbenzoic acid

  • MDL number MFCD09835285
  • Cas Number : 367954-99-2
  • Molecular Weight : 172.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-difluoro-5-methylbenzoic-acid

Safety Information

MSDS https://store.apolloscientific.co.uk/product/24-difluoro-5-methylbenzoic-acid
Availability details for
Quantity
  • Availability
  • Add to Cart