APOH03871DE7 Apollo Scientific Ltd

3-Chloro-7-(trifluoromethyl)-1H-indazole

  • Cas Number : 1388070-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=111323

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=111323
Availability details for
Quantity
  • Availability
  • Add to Cart