APOH03871DFB Apollo Scientific Ltd

1-(3-Chloro-4-fluorophenyl)-3-(dimethylamino)prop-2-en-1-one

  • Cas Number : 154258-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=111372

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=111372
Availability details for
Quantity
  • Availability
  • Add to Cart