APOH03871E29 Apollo Scientific Ltd

2-Chloro-5-fluoro-1-methyl-1H-indole-3-carbaldehyde

  • Cas Number : 1310141-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=111643

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=111643
Availability details for
Quantity
  • Availability
  • Add to Cart