APOH03871E2B Apollo Scientific Ltd

Ethyl 2-bromo-4-fluorobenzoate

  • MDL number MFCD06204924
  • Cas Number : 651341-68-3
  • Molecular Weight : 247.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-bromo-4-fluorobenzoate

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-bromo-4-fluorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart