APOH03871E2F Apollo Scientific Ltd

Ethyl 2,4-difluoro-5-iodobenzoate

  • Cas Number : 1541937-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=111662

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=111662
Availability details for
Quantity
  • Availability
  • Add to Cart