APOH03871E34 Apollo Scientific Ltd

3-Chloro-1H-indole-2-carboxylic acid

  • MDL number MFCD11007738
  • Cas Number : 28737-32-8
  • Molecular Weight : 195.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-1h-indole-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-1h-indole-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart