APOH03872950 Apollo Scientific Ltd

1,4-Oxathiane-2,6-dione

  • MDL number MFCD00051689
  • Cas Number : 3261-87-8
  • Molecular Weight : 132.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-98
Synonym Thiodiglycolic anhydride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/14-oxathiane-26-dione

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/14-oxathiane-26-dione
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart