APOH0387295D Apollo Scientific Ltd

Methyl 3-formyl-2-nitrobenzoate

  • MDL number MFCD14706049
  • Cas Number : 138229-59-1
  • Molecular Weight : 209.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Flash Point 173
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 354
Product Link https://store.apolloscientific.co.uk/product/methyl-3-formyl-2-nitrobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-formyl-2-nitrobenzoate
pictograms GHS07
hstatements H302;H315;H335;H412
Availability details for
Quantity
  • Availability
  • Add to Cart