APOH03872971 Apollo Scientific Ltd

2-Chloro-6-fluoro-1,3-benzoxazole

  • MDL number MFCD16250625
  • Cas Number : 153403-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-6-fluorobenzo[d]oxazole
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-fluoro-13-benzoxazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-fluoro-13-benzoxazole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart