APOH039F93B5 Apollo Scientific Ltd

1-(2-Bromoethynyl)-2-fluorobenzene

  • MDL number MFCD31536744
  • Cas Number : 1246224-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=112183

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=112183
pictograms GHS05;GHS07
hstatements H302+H312+H332;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart