APOH039F93C5 Apollo Scientific Ltd

N-Methylbicyclo[1.1.1]pentan-1-amine hydrochloride

  • MDL number MFCD31417451
  • Cas Number : 1882089-75-9
  • Molecular Weight : 133.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-methylbicyclo[1.1.1]pentan-1-amine hydrochloride
Inchi Code 1S/C6H11N.ClH/c1-7-6-2-5(3-6)4-6;/h5,7H,2-4H2,1H3;1H
Inchi Key NRZPQUZMLISKHJ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-methylbicyclo111pentan-1-amine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n-methylbicyclo111pentan-1-amine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart