APOH039F93CB Apollo Scientific Ltd

Methyl 5-chloro-2-methyl-1H-indole-3-carboxylate

  • MDL number MFCD23159660
  • Cas Number : 762288-06-2
  • Molecular Weight : 223.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-chloro-2-methyl-1H-indole-3-carboxylate
Inchi Code 1S/C11H10ClNO2/c1-6-10(11(14)15-2)8-5-7(12)3-4-9(8)13-6/h3-5,13H,1-2H3
Inchi Key KIDKWAXCVMNPDL-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-chloro-2-methyl-1h-indole-3-carboxylate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-chloro-2-methyl-1h-indole-3-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart