APOH039F93DA Apollo Scientific Ltd

3-Iodo-7-methoxy-1H-pyrrolo[2,3-c]pyridine

  • MDL number MFCD12963282
  • Cas Number : 1190316-96-1
  • Molecular Weight : 274.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-iodo-7-methoxy-1H-pyrrolo[2,3-c]pyridine
Inchi Code 1S/C8H7IN2O/c1-12-8-7-5(2-3-10-8)6(9)4-11-7/h2-4,11H,1H3
Inchi Key RNQCGSZKLYMHCD-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-iodo-7-methoxy-1h-pyrrolo23-cpyridine

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-iodo-7-methoxy-1h-pyrrolo23-cpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart