APOH039F93DC Apollo Scientific Ltd

5-Formylpyridine-2-carbonitrile

  • MDL number MFCD12755972
  • Cas Number : 131747-68-7
  • Molecular Weight : 132.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-formyl-2-pyridinecarbonitrile
Inchi Code 1S/C7H4N2O/c8-3-7-2-1-6(5-10)4-9-7/h1-2,4-5H
Inchi Key ZYOAZOZQOTWMEE-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-formylpyridine-2-carbonitrile

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-formylpyridine-2-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart