APOH039F93DE Apollo Scientific Ltd

5-(tert-Butyldimethylsilyloxy)-2-fluorobenzaldehyde

  • MDL number MFCD17676576
  • Cas Number : 113984-67-1
  • Molecular Weight : 254.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-{[tert-butyl(dimethyl)silyl]oxy}-2-fluorobenzaldehyde
Inchi Code 1S/C13H19FO2Si/c1-13(2,3)17(4,5)16-11-6-7-12(14)10(8-11)9-15/h6-9H,1-5H3
Inchi Key LCRGCKGCZUAUDG-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-tert-butyldimethylsilyloxy-2-fluorobenzaldehyde

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-tert-butyldimethylsilyloxy-2-fluorobenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart