APOH039F93E2 Apollo Scientific Ltd

4-Bromo-1-methyl-1H-indole-6-carboxylic acid

  • MDL number MFCD31536723
  • Cas Number : 1782217-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=112206

Safety Information

signalword Warning
pcodes P280;P304+P340;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=112206
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart